Invitrogen
Manufacturer Code:PA5115225
Catalog # PIPA5115225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FGF1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acidic eye-derived fibroblast growth factor acidic eye-derived growth factor II Acidic fibroblast growth factor aFGF beta-endothelial cell growth factor Dffrx ECGF ECGFA ECGFB ECGF-beta EDGF EDGF II Endothelial cell growth factor Endothelial cell growth factor alpha Endothelial cell growth factor beta endothelial cell growth factor beta and alpha chains endothelial cell growth factor alpha endothelial cell growth factor beta endothelial growth factor Fam FGF FGF1 Fgf-1 FGF1 protein Fgfa FGF-alpha Fibroblast growth factor fibroblast growth factor 1 fibroblast growth factor 1 (acidic) Fibroblast growth factor 1 (heparin binding) fibroblast growth factor acidic GLIO703 HBGF1 HBGF-1 heparin-binding growth factor heparin-binding growth factor 1 H-FGF-a M-FGF-a Prostatropin unnamed protein product; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; ECGF; Endothelial cell growth factor; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; FGF-1; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); HBGF-1; Heparin-binding growth factor 1
Gene Aliases: AFGF; Dffrx; ECGF; ECGF-beta; ECGFA; ECGFB; Fam; FGF-1; FGF-alpha; FGF1; FGFA; GLIO703; HBGF-1; HBGF1
UniProt ID: (Human) P05230, (Mouse) P61148
Entrez Gene ID: (Human) 2246, (Mouse) 14164
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.