Novus Biologicals
Manufacturer Code:NB600778C
Catalog # NB600778C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MC-GF1 |
Immunogen | Recombinant full length protein (Human). |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Basic fibroblast growth factor; Basic fibroblast growth factor basic fibroblast growth factor bFGF bFGF FGFBprostatropin fibroblast growth factor 2 (basic) HBGF-2 heparin-binding growth factor 2; basic fibroblast growth factor bFGF; bFGF; FGF-2; Fibroblast growth factor 2; fibroblast growth factor 2 (basic); HBGF-2; Heparin-binding growth factor 2; prostatropin
Gene Aliases: BFGF; FGF-2; FGF2; FGFB; HBGF-2
UniProt ID: (Human) P09038
Entrez Gene ID: (Human) 2247
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.