Novus Biologicals
Manufacturer Code:NBP212305V
Catalog # NBP212305V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0291-8D24 |
Immunogen | Human FGF8 |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AIGF; AIGFKAL6 Androgen-induced growth factor FGF-8 fibroblast growth factor 8 fibroblast growth factor 8 (androgen-induced) HBGF-8 Heparin-binding growth factor 8 MGC149376; Androgen-induced growth factor; FGF-8; Fibroblast growth factor 8; fibroblast growth factor 8 (androgen-induced); HBGF-8; Heparin-binding growth factor 8
Gene Aliases: AIGF; FGF-8; FGF8; HBGF-8; HH6; KAL6
UniProt ID: (Human) P55075
Entrez Gene ID: (Human) 2253
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.