R&D Systems
Manufacturer Code:AF423SP
Catalog # AF423SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse FGF-8b Gln23-Arg215 Accession # NP_006110 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AIGF; AIGF AIGFKAL6 Androgen-induced growth factor FGF8 FGF-8 fibroblast growth factor 8 fibroblast growth factor 8 (androgen-induced) HBGF-8 Heparin-binding growth factor 8 MGC149376; Androgen-induced growth factor; FGF-8; Fibroblast growth factor 8; fibroblast growth factor 8 (androgen-induced); HBGF-8; Heparin-binding growth factor 8
Gene Aliases: AIGF; FGF-8; FGF8; HBGF-8; HH6; KAL6
UniProt ID: (Human) P55075
Entrez Gene ID: (Human) 2253
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.