Novus Biologicals
Manufacturer Code:NBP237374
Catalog # NBP237374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A12G9 |
Immunogen | Purified recombinant fragment of humanFGF4 (AA: 62-123) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FGF-4; Fibroblast growth factor 4; fibroblast growth factor 4 HBGF-4Transforming protein KS3 heparin secretory transforming protein 1 Heparin secretory-transforming protein 1 Heparin-binding growth factor 4 HST-1HSTF-1 HSTF1fibroblast growth factor 4 splice isoform HSTFGF-4 human stomach cancer transforming factor from FGF-related oncogene kaposi sarcoma oncogene KFGF K-FGF KS3 oncogene HST; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; Heparin secretory-transforming protein 1; Heparin-binding growth factor 4; HST; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; oncogene HST; Transforming protein KS3
Gene Aliases: FGF4; HBGF-4; HST; HST-1; HSTF1; K-FGF; KFGF; KS3
UniProt ID: (Human) P08620
Entrez Gene ID: (Human) 2249
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.