Novus Biologicals
Manufacturer Code:NBP212297B
Catalog # NBP212297B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0283-6G2 |
Immunogen | Human FGF16 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FGF-16; FGF-16 fibroblast growth factor 16; Fibroblast growth factor 16; metacarpal 4-5 fusion
Gene Aliases: FGF-16; FGF16; MF4
UniProt ID: (Human) O43320
Entrez Gene ID: (Human) 8823
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.