Novus Biologicals
Manufacturer Code:NBP270721JF549
Catalog # N270721J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7E8 |
Immunogen | Full length recombinant protein of human FGF-13 (NP_004105) produced in E.coli. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FGF-13; FGF-13 FGF2Fibroblast growth factor homologous factor 2 FHF2FHF-2 fibroblast growth factor 13; FHF-2; Fibroblast growth factor 13; Fibroblast growth factor homologous factor 2
Gene Aliases: FGF-13; FGF13; FGF2; FHF-2; FHF2
UniProt ID: (Human) Q92913
Entrez Gene ID: (Human) 2258
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.