Novus Biologicals
Manufacturer Code:NBP212295AF405
Catalog # NB212295405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0281-6L27 |
Immunogen | Human FGF10 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FGF-10; FGF-10 fibroblast growth factor 10 Keratinocyte growth factor 2 produced by fibroblasts of urinary bladder lamina propria; Fibroblast growth factor 10; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria
Gene Aliases: FGF10
UniProt ID: (Human) O15520
Entrez Gene ID: (Human) 2255
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.