R&D Systems
Manufacturer Code:FAB10082S100UG
Catalog # FB10082S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1000624 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human FFAR2/GPR43 Met1-Glu330 Accession # O15552 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: fatty acid receptor 2; free fatty acid activated receptor 2; free fatty acid activated receptor 2 free fatty acid receptor 2 G protein-coupled receptor 43 GPR43 GPR43FFA2R G-protein coupled receptor 43; Free fatty acid receptor 2; G protein-coupled receptor 43; G-protein coupled receptor 43
Gene Aliases: FFA2; FFA2R; FFAR2; GPCR43; GPR43
UniProt ID: (Human) O15552
Entrez Gene ID: (Human) 2867
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.