Proteintech
Manufacturer Code:199521AP150UL
Catalog # 50-173-2619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fatty acid receptor 2; FFA2R FFAR2 free fatty acid receptor 2 G protein coupled receptor 43 GPR43; free fatty acid activated receptor 2; Free fatty acid receptor 2; G protein-coupled receptor 43; G-protein coupled receptor 43
Gene Aliases: FFA2; FFA2R; FFAR2; GPCR43; GPR43
UniProt ID: (Human) O15552
Entrez Gene ID: (Pig) 100126285, (Human) 2867
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.