Invitrogen
Manufacturer Code:PA543562
Catalog # PIPA543562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FEZF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FEZ family Zinc finger 1 ZNF312B FEZ; Fez family zinc finger protein 1; Zinc finger protein 312B
Gene Aliases: FEZ; FEZF1; HH22; RGD1560480; ZNF312B
UniProt ID: (Human) A0PJY2
Entrez Gene ID: (Horse) 100071453, (Rabbit) 100351321, (Human) 389549, (Rat) 500048, (Guinea pig) 100722372
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.