Invitrogen
Manufacturer Code:PA530928
Catalog # PIPA530928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 315 and 596 of Human FEZ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F37 FEZ1 F37/Esophageal cancer-related gene-coding leucine-zipper motif leucine zipper putative tumor suppressor 1; fasciculation and elongation protein zeta 1 (zygin I); Fasciculation and elongation protein zeta-1; Zygin I; Zygin-1
Gene Aliases: FEZ1; UNC-76
UniProt ID: (Human) Q99689
Entrez Gene ID: (Human) 9638
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.