Novus Biologicals
Manufacturer Code:NBP176395
Catalog # NBP176395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human FEZ1. The immunogen is located within amino acids 270 - 320 of FEZ1. Amino Acid Squence: KQKEQRELMKKRRKEKGL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fasciculation and elongation protein zeta 1 (zygin I); fasciculation and elongation protein zeta 1 (zygin I) fasciculation and elongation protein zeta-1 Zygin I zygin-1; Fasciculation and elongation protein zeta-1; Zygin I; Zygin-1
Gene Aliases: FEZ1; UNC-76
UniProt ID: (Human) Q99689
Entrez Gene ID: (Human) 9638
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.