Abnova
Manufacturer Code:PAB7151
Catalog # 89-132-995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human FEZ1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of FEZ1.
For Research Use Only
Protein Aliases: fasciculation and elongation protein zeta 1 (zygin I); Fasciculation and elongation protein zeta-1; Zygin I; Zygin-1
Gene Aliases: FEZ1; UNC-76
UniProt ID: (Human) Q99689
Entrez Gene ID: (Human) 9638
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.