Invitrogen
Manufacturer Code:PA595067
Catalog # PIPA595067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human FES identical to the related mouse and rat sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: feline sarcoma (Snyder-Theilen) viral (v-fes)/Fujinami avian sarcoma (PRCII) viral (v-fps) oncogene homolog; feline sarcoma oncogene; Feline sarcoma/Fujinami avian sarcoma oncogene homolog; Oncogene FES, feline sarcoma virus; p93c-fes; Proto-oncogene c-Fes; Proto-oncogene c-Fps; proto-oncogene tyrosine-protein kinase Fes/Fps; tyrosine kinase Fps/Fes; Tyrosine-protein kinase Fes/Fps; Tyrosine-protein kinase Fes/Fps 2.7.10.2 Feline sarcoma/Fujinami avian sarcoma oncogene homolog Proto-oncogene c-Fes Proto-oncogene c-Fps p93c-fes FES FPS
Gene Aliases: AI586313; BB137047; c-fes; FES; FPS; RGD1564385
UniProt ID: (Human) P07332, (Mouse) P16879
Entrez Gene ID: (Human) 2242, (Mouse) 14159, (Rat) 361597
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.