Abnova
Manufacturer Code:PAB16550
Catalog # 89-118-419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 19 amino acids of human FERMT3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of FERMT3.
For Research Use Only
Protein Aliases: Fermitin family homolog 3; KIND3; kindlin 3; Kindlin-3; MGC10966; MIG-2; MIG2-like protein; MIG2B; UNC-112 related protein 2; Unc-112-related protein 2; UNC112C; URP2; URP2SF
Gene Aliases: FERMT3; KIND3; MIG-2; MIG2B; UNC112C; URP2; URP2SF
UniProt ID: (Human) Q8N207
Entrez Gene ID: (Human) 83706
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.