Proteintech
Manufacturer Code:181311AP150UL
Catalog # 50-173-2003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FERMT3 Fusion Protein Ag12713 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fermitin family homolog 3; Fermitin family homolog 3 FERMT3 KIND3 Kindlin 3 MIG 2 MIG2 like protein MIG2B Unc 112 related protein 2 UNC112C URP2 URP2SF; kindlin 3; Kindlin-3; MIG2-like protein; UNC-112 related protein 2; Unc-112-related protein 2
Gene Aliases: C79673; FERMT3; KIND3; Kindlin3; MIG-2; MIG2B; RGD1310168; UNC112C; URP2; URP2SF
UniProt ID: (Human) Q8N207, (Mouse) Q8K1B8
Entrez Gene ID: (Human) 83706, (Mouse) 108101, (Rat) 309186
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.