OriGene
Manufacturer Code:CF500505
Catalog # 50-165-9575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI14A11 |
Immunogen | Full-length protein expressed in 293T cell transfected with human FERMT2 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Fermitin family homolog 2; KIND2 MIG2 PLEKHC1 UNC112 UNC112B mig-2; kindlin 2; Kindlin-2; MIG-2; mitogen inducible gene 2 protein; Mitogen-inducible gene 2 protein; PH domain-containing family C member 1; pleckstrin homology domain containing, family C (with FERM domain) member 1; pleckstrin homology domain containing, family C member 1; Pleckstrin homology domain-containing family C member 1
Gene Aliases: FERMT2; KIND2; mig-2; MIG2; PLEKHC1; UNC112; UNC112B
UniProt ID: (Human) Q96AC1
Entrez Gene ID: (Human) 10979, (Dog) 490693, (Rat) 289992
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.