Novus Biologicals
Manufacturer Code:NBP180169
Catalog # NBP180169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human FERD3L. Peptide sequence SCVDTTVLDFVADLSLASPRRPLLCDFAPGVSLGDPALALREGRPRRMAR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic helix-loop-helix protein N-twist; Basic helix-loop-helix protein N-twist BHLHA31 bHLHa31NTWIST Class A basic helix-loop-helix protein 31 Fer3-like (Drosophila) fer3-like protein MGC119861 NATO3Neuronal twist Nephew of atonal 3 N-TWIST; bHLHa31; Class A basic helix-loop-helix protein 31; Fer3-like protein; Nephew of atonal 3; Neuronal twist
Gene Aliases: BHLHA31; FERD3L; N-TWIST; NATO3; NTWIST
UniProt ID: (Human) Q96RJ6
Entrez Gene ID: (Human) 222894
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.