Invitrogen
Manufacturer Code:MA524734
Catalog # PIMA524734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1487CT794.8.50 |
Immunogen | Mouse Fer recombit protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: fer (fms/fps related) protein kinase, testis specific 2; p94-Fer; Proto-oncogene c-Fer; proto-oncogene tyrosine-protein kinase Fer; Tyrosine-protein kinase Fer; Tyrosine-protein kinase Fer Proto-oncogene c-Fer p94-Fer Fer Fert2
Gene Aliases: AV082135; C330004K01Rik; Fer; Fert; Fert2
UniProt ID: (Mouse) P70451
Entrez Gene ID: (Mouse) 14158
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.