Invitrogen
Manufacturer Code:PA579248
Catalog # PIPA579248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human FER. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-FER; Feline encephalitis virus-related kinase FER; fer (fms/fps related) protein kinase, testis specific 2; fer (fps/fes related) tyrosine kinase; Fujinami poultry sarcoma/Feline sarcoma-related protein Fer; p94-FER; phosphoprotein NCP94; protein phosphatase 1, regulatory subunit 74; Proto-oncogene c-Fer; proto-oncogene tyrosine-protein kinase FER; Tyrosine kinase 3; tyrosine protein kinase FLK; Tyrosine-protein kinase Fer; Tyrosine-protein kinase Fer Feline encephalitis virus-related kinase FER Fujinami poultry sarcoma/Feline sarcoma-related protein Fer Proto-oncogene c-Fer Tyrosine kinase 3 p94-Fer FER TYK3; Tyrosine-protein kinase FLK
Gene Aliases: FER; Fert2; Flk; Flk_retired; p94-Fer; PPP1R74; TYK3
UniProt ID: (Human) P16591, (Rat) P09760
Entrez Gene ID: (Human) 2241, (Rat) 301737
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.