Novus Biologicals
Manufacturer Code:NBP270716AF750
Catalog # NP270716A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F3 |
Immunogen | Recombinant protein fragment corresponding to amino acids 117-380 of human FEN-1 (NP_004102) produced in E.coli. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DNase IV; EC 3.1 FEN-1hFEN-1 flap structure-specific endonuclease 1DNase IV Maturation factor 1 maturation factor-1 MF1flap endonuclease 1 RAD2; FEN-1; FEN1; Flap endonuclease 1; Flap structure-specific endonuclease 1; Maturation factor 1; maturation factor-1; MF1
Gene Aliases: FEN-1; FEN1; MF1; RAD2
UniProt ID: (Human) P39748
Entrez Gene ID: (Human) 2237
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.