Novus Biologicals
Manufacturer Code:NBP298793B
Catalog # NB098450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human FEN-1. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNase IV; EC 3.1 FEN-1hFEN-1 flap structure-specific endonuclease 1DNase IV Maturation factor 1 maturation factor-1 MF1flap endonuclease 1 RAD2; FEN-1; FEN1; Flap endonuclease 1; Flap structure-specific endonuclease 1; Maturation factor 1; maturation factor-1; MF1
Gene Aliases: FEN-1; FEN1; MF1; RAD2
UniProt ID: (Human) P39748
Entrez Gene ID: (Human) 2237
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.