Invitrogen
Manufacturer Code:PA517432
Catalog # PIPA517432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues of human FE65 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adaptor protein FE65a2; adaptor protein FE65a2 amyloid beta A4 precursor protein-binding family B member 1 amyloid beta A4 precursor protein-binding family B member 1 stat-like protein; amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65); amyloid beta A4 precursor protein-binding family B member 1; Amyloid-beta A4 precursor protein-binding family B member 1; Protein Fe65; stat-like protein
Gene Aliases: APBB1; FE65; MGC:9072; RIR
UniProt ID: (Human) O00213, (Rat) P46933, (Mouse) Q9QXJ1
Entrez Gene ID: (Human) 322, (Rat) 29722, (Mouse) 11785
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.