Novus Biologicals
Manufacturer Code:NBP299557AF532
Catalog # NB085673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human FE65. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adaptor protein FE65a2; adaptor protein FE65a2 amyloid beta (A4) precursor protein-binding family B member 1 (Fe65) amyloid beta A4 precursor protein-binding family B member 1 Fe65 FE65amyloid beta A4 precursor protein-binding family B member 1 MGC:9072 Protein Fe65 RIR stat-like protein; amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65); Amyloid-beta A4 precursor protein-binding family B member 1; Protein Fe65; stat-like protein
Gene Aliases: APBB1; FE65; MGC:9072; RIR
UniProt ID: (Human) O00213
Entrez Gene ID: (Human) 322
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.