Invitrogen
Manufacturer Code:PA596260
Catalog # PIPA596260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 252-491 of human FDXR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenodoxin reductase; adrenodoxin-NADP(+) reductase; AR; Ferredoxin reductase; Ferredoxin--NADP(+) reductase; NADPH:adrenodoxin oxidoreductase, mitochondrial
Gene Aliases: ADXR; AR; FDXR
UniProt ID: (Human) P22570, (Rat) P56522, (Mouse) Q61578
Entrez Gene ID: (Human) 2232, (Rat) 79122, (Mouse) 14149
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.