Proteintech
Manufacturer Code:249811AP150UL
Catalog # 50-173-4653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FDXACB1 Fusion Protein Ag21785 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FDX-ACDB domain-containing protein 1; FDXACB1; Ferredoxin-fold anticodon-binding domain-containing protein 1; hypothetical protein BC006136
Gene Aliases: FDXACB1; hCG_2033039
UniProt ID: (Human) Q9BRP7
Entrez Gene ID: (Human) 91893
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.