Invitrogen
Manufacturer Code:PA581388
Catalog # PIPA581388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse FDPS/Farnesyl Diphosphate Synthase protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (2E,6E)-farnesyl diphosphate synthase; Cholesterol-regulated 39 kDa protein; CR 39; Dimethylallyltranstransferase; Farnesyl diphosphate synthase; Farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; FPP synthase; FPP synthetase; FPS; Geranyltranstransferase
Gene Aliases: 6030492I17Rik; AI256750; Fdps; Fdpsl1; mKIAA1293
UniProt ID: (Mouse) Q920E5
Entrez Gene ID: (Mouse) 110196
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.