Proteintech
Manufacturer Code:161291AP150UL
Catalog # 50-173-0738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FDPS Fusion Protein Ag9115 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (2E,6E)-farnesyl diphosphate synthase; Cholesterol-regulated 39 kDa protein; CR 39; Dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase); farnesyl diphosphate synthetase; Farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; FDPS FPP synthase FPPS FPS KIAA1293; FPP synthase; FPP synthetase; FPS; Geranyltranstransferase; testis-specific farnesyl pyrophosphate synthetase
Gene Aliases: 6030492I17Rik; Ac2-125; AI256750; FDPS; Fdpsl1; FPPS; FPS; KIAA1293; mKIAA1293; POROK9
UniProt ID: (Human) P14324, (Rat) P05369, (Mouse) Q920E5
Entrez Gene ID: (Human) 2224, (Rat) 83791, (Mouse) 110196
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.