Invitrogen
Manufacturer Code:PA544284
Catalog # PIPA544284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FCHO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-BAR domain only protein 1; FCH domain only 1; FCH domain only protein 1; unm_sa2424
Gene Aliases: 3322402E17Rik; FCHO1; KIAA0290; N28152
UniProt ID: (Human) O14526, (Mouse) Q8K285
Entrez Gene ID: (Dog) 476673, (Human) 23149, (Rat) 290639, (Mouse) 74015, (Zebrafish) 100913078
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.