Proteintech
Manufacturer Code:267671AP150UL
Catalog # 50-173-5450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FCHO1 Fusion Protein Ag25291 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-BAR domain only protein 1; FCH domain only 1 FCH domain only protein 1 FCHO1 KIAA0290; FCH domain only protein 1
Gene Aliases: FCHO1; KIAA0290
UniProt ID: (Human) O14526
Entrez Gene ID: (Human) 23149
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.