Invitrogen
Manufacturer Code:PA547871
Catalog # PIPA547871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived mouse FCRN Ser22-Ser301 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FCGRT alpha-chain Fc fragment of IgG receptor transporter alpha FcRn FcRn alpha chain FCRNimmunoglobulin receptor intestinal heavy chain IgG Fc fragment receptor transporter alpha chain IgG receptor FcRn large subunit p51 major histocompatibility complex class I-like Fc receptor Neonatal Fc receptor neonatal Fc-receptor for Ig; FcRn; IgG Fc fragment receptor transporter alpha chain; IgG receptor FcRn large subunit p51; Neonatal Fc receptor
Gene Aliases: Fcgrt; FcRn
UniProt ID: (Mouse) Q61559
Entrez Gene ID: (Mouse) 14132
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.