Invitrogen
Manufacturer Code:PA5115222
Catalog # PIPA5115222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human FCER1G corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI573376 CD23 Fc epsilon receptor Ig Fc fragment of IgE receptor Ig Fc fragment of IgE high affinity I receptor for gamma polypeptide Fc receptor gamma-chain Fc receptor IgE high affinity I gamma polypeptide Fce1g fc-epsilon RI-gamma FcepsilonRI FCER1G fceRI gamma FcR[g] FCRG FcRgamma FcR-gamma high affinity immunoglobulin epsilon receptor subunit gamma IgE Fc receptor subunit gamma immunoglobulin E receptor high affinity gamma chain Ly-50; Fc epsilon receptor Ig; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc receptor, IgE, high affinity I, gamma polypeptide; Fc-epsilon RI-gamma; FceRI gamma; FcRgamma; gamma polypeptide; High affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain
Gene Aliases: AI573376; CD23; Fce1g; FcepsilonRI; FCER1G; FcR-gamma; FcR[g]; FCRG; FcRgamma; Ly-50
UniProt ID: (Human) P30273, (Rat) P20411, (Mouse) P20491
Entrez Gene ID: (Human) 2207, (Rat) 25441, (Mouse) 14127
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.