Invitrogen
Manufacturer Code:PA5109716
Catalog # PIPA5109716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 19-86 of human FCER1G (NP_004097.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI573376 CD23 Fc receptor gamma-chain Fce1g Fc-epsilon RI-gamma FcepsilonRI Fcer1g FceRI gamma FcR[g] FCRG FcRgamma FcR-gamma High affinity immunoglobulin epsilon receptor subunit gamma IgE Fc receptor subunit gamma Ly-50; Fc epsilon receptor Ig; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc-epsilon RI-gamma; FceRI gamma; FcRgamma; High affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain
Gene Aliases: AI573376; CD23; Fce1g; FcepsilonRI; FCER1G; FcR-gamma; FcR[g]; FCRG; FcRgamma; Ly-50
UniProt ID: (Human) P30273, (Mouse) P20491
Entrez Gene ID: (Human) 2207, (Mouse) 14127
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.