Novus Biologicals
Manufacturer Code:NBP2849300.1ML
Catalog # NB032859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human FCER1G |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fc epsilon receptor Ig; Fc fragment of IgE high affinity I receptor for gamma polypeptide Fc-epsilon RI-gamma FceRI gamma FCRG high affinity immunoglobulin epsilon receptor subunit gamma IgE Fc receptor subunit gamma immunoglobulin E receptor high affinity gamma chain; Fc fragment of IgE, high affinity I, receptor for; gamma polypeptide; Fc receptor gamma-chain; Fc-epsilon RI-gamma; FceRI gamma; FcRgamma; High affinity immunoglobulin epsilon receptor subunit gamma; IgE Fc receptor subunit gamma; immunoglobulin E receptor, high affinity, gamma chain
Gene Aliases: FCER1G; FCRG
UniProt ID: (Human) P30273
Entrez Gene ID: (Human) 2207
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.