Novus Biologicals
Manufacturer Code:NBP290841AF594
Catalog # NB078747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 13 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Rat FCAR/CD89 (NP_973721.1 Met1-Asn228). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD89; CD89 antigen CD89IgA Fc receptor Fc alpha receptor Fc fragment of IgA receptor for immunoglobulin alpha Fc receptor; Fc alpha receptor; Fc fragment of IgA, receptor for; IgA Fc receptor; Immunoglobulin alpha Fc receptor
Gene Aliases: CD89; CTB-61M7.2; FcalphaRI; FCAR
UniProt ID: (Human) Q13603
Entrez Gene ID: (Human) 2204
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.