Abnova
Manufacturer Code:MAB1660
Catalog # 89-110-556
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MIP7c |
Immunogen | Recombinant protein corresponding to extracellular reagion of human FCAR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant FCAR.
For Research Use Only
Protein Aliases: CD89; Fc alpha receptor; Fc fragment of IgA, receptor for; IgA Fc receptor; Immunoglobulin alpha Fc receptor
Gene Aliases: CD89; CTB-61M7.2; FcalphaRI; FCAR
UniProt ID: (Human) Q13603
Entrez Gene ID: (Human) 2204
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.