Invitrogen
Manufacturer Code:PA544382
Catalog # PIPA544382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FBXW8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-box and WD repeat domain containing 8 FBW6 FBW8 FBX29 FBXO29 MGC33534 FBXW6; F-box and WD-40 domain protein 8; F-box and WD-40 domain-containing protein 8; F-box only protein 29; F-box protein FBX29; F-box/WD repeat-containing protein 8
Gene Aliases: 4930438M06Rik; FBW6; FBW8; FBX29; FBXO29; FBXW6; FBXW8
UniProt ID: (Human) Q8N3Y1, (Mouse) Q8BIA4
Entrez Gene ID: (Dog) 106557503, (Rabbit) 100345880, (Human) 26259, (Bovine) 787118, (Horse) 100055213, (Rat) 304522, (Mouse) 231672, (Guinea pig) 100728842
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.