Novus Biologicals
Manufacturer Code:NBP184715
Catalog # NBP184715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KLIFQECRAKEHMLQTNWKNRKGAVSELEHVPDTVLCDVHSHDGVVIAGYTSGDVRVWDTRTWDYVAPFLESEDEEDEPGMQPNVSFVRINSSLAVAA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-box and WD repeat domain containing 8 F-box only protein 29F-box/WD repeat-containing protein 8 FBW8F-box and WD-40 domain-containing protein 8 FBX29MGC33534 FBXO29 FBXW6FBW6F-box and WD-40 domain protein 8; F-box and WD-40 domain protein 8; F-box and WD-40 domain-containing protein 8; F-box only protein 29; F-box/WD repeat-containing protein 8
Gene Aliases: FBW6; FBW8; FBX29; FBXO29; FBXW6; FBXW8
UniProt ID: (Human) Q8N3Y1
Entrez Gene ID: (Human) 26259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.