Novus Biologicals
Manufacturer Code:NBP159631
Catalog # NBP159631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to FBXW7(F-box and WD repeat domain containing 7) The peptide sequence was selected from the C terminal of FBXW7 (NP_001013433). Peptide sequence LKTGEFIRNLVTLESGGSGGVVWRIRASNTKLVCAVGSRNGTEETKLLVL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGOFBXO30 Archipelago homolog archipelago Drosophila homolog of CDC4FLJ16457 F-box and WD repeat domain containing 7 F-box and WD-40 domain protein 7 (archipelago homolog Drosophila) F-box and WD-40 domain-containing protein 7 F-box protein FBW7 F-box protein FBX30 F-box protein SEL-10 F-box/WD repeat-containing protein 7 FBW7hCdc4 FBX30FLJ11071 FBXW6 hAgo homolog of C elegans sel-10 SEL-10DKFZp686F23254 SEL10FBW6; archipelago; Archipelago homolog; F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase; F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila); F-box and WD-40 domain-containing protein 7; F-box protein FBW7; F-box protein FBX30; F-box protein SEL-10; F-box/WD repeat-containing protein 7; hAgo; hCdc4; homolog of C elegans sel-10; SEL-10
Gene Aliases: AGO; CDC4; FBW6; FBW7; FBX30; FBXO30; FBXW6; FBXW7; hAgo; hCdc4; SEL-10; SEL10
UniProt ID: (Human) Q969H0
Entrez Gene ID: (Human) 55294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.