Invitrogen
Manufacturer Code:PA542300
Catalog # PIPA542300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FBXO7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-box only protein 7; F-box protein 7 DKFZp686B08113 FBX FBX07 FBX7 PKPS PARK15
Gene Aliases: 2410015K21Rik; A230052G17Rik; AI117679; AW260462; C85118; FBX; FBX07; FBX7; FBXO7; PARK15; PKPS; si:ch211-286m4.2; zgc:114096
UniProt ID: (Bovine) Q2T9S7, (Human) Q9UKT2, (Mouse) Q3U7U3, (Rat) Q68FS3
Entrez Gene ID: (Dog) 474522, (Pig) 100155281, (Rabbit) 100328739, (Bovine) 508235, (Horse) 100068847, (Human) 25793, (Guinea pig) 100735044, (Mouse) 69754, (Rat) 366854, (Zebrafish) 558097
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.