Novus Biologicals
Manufacturer Code:NBP297739JF646
Catalog # NB047260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human FBXO7 fragment. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-box only protein 7; F-box only protein 7 F-box protein 7 FBX FBX07 FBX7Fbx PARK15DKFZp686B08113 PKPS
Gene Aliases: FBX; FBX07; FBX7; FBXO7; PARK15; PKPS
UniProt ID: (Human) Q9UKT2
Entrez Gene ID: (Human) 25793
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.