Proteintech
Manufacturer Code:106961AP150UL
Catalog # 50-172-6521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FBXO7 Fusion Protein Ag1035 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F box only protein 7 F box protein 7 FBX FBX07 FBX7 FBXO7 PARK15 PKPS; F-box only protein 7
Gene Aliases: 2410015K21Rik; A230052G17Rik; AI117679; AW260462; C85118; FBX; FBX07; FBX7; FBXO7; PARK15; PKPS
UniProt ID: (Human) Q9UKT2, (Mouse) Q3U7U3
Entrez Gene ID: (Human) 25793, (Mouse) 69754
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.