Abnova
Manufacturer Code:MAB13276
Catalog # 89-934-124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | EMI1/1176 |
Immunogen | Recombinant protein corresponding to 203 residues around amino acids 1-250 of human FBXO5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant human FBXO5.
For Research Use Only
Protein Aliases: Early mitotic inhibitor 1; EMI1; F-box only protein 5; F-box protein Fbx5; FBX5; Fbxo31
Gene Aliases: EMI1; FBX5; Fbxo31; FBXO5
UniProt ID: (Human) Q9UKT4
Entrez Gene ID: (Human) 26271
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.