Novus Biologicals
Manufacturer Code:NBP2827650.1ML
Catalog # NB030694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human FBXO45 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-box only protein 45; F-box only protein 45 F-box protein 45 F-box/SPRY domain-containing protein 1 Fbx45; F-box/SPRY domain-containing protein 1; hFbxo45
Gene Aliases: FBX45; FBXO45
UniProt ID: (Human) P0C2W1
Entrez Gene ID: (Human) 200933
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.