Proteintech
Manufacturer Code:145901AP150UL
Catalog # 50-172-9565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FBXO2 Fusion Protein Ag6122 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F box only protein 2 F box protein 2 FBG1 Fbs1 FBX2 FBXO2 NFB42; F-box gene 1; F-box only protein 2; neural F box protein NFB42; organ of Corti protein 1; prion protein ligand 4
Gene Aliases: FBG1; Fbs1; Fbs2; FBX2; FBXO2; NFB42; OCP1; Prpl4
UniProt ID: (Human) Q9UK22, (Mouse) Q80UW2
Entrez Gene ID: (Human) 26232, (Mouse) 230904, (Rat) 85273
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.