Abnova
Manufacturer Code:PAB20133
Catalog # 89-121-635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human FBXO18. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant FBXO18.
For Research Use Only
Protein Aliases: F-box DNA helicase 1; F-box only protein 18; F-box only protein, helicase, 18; FBH1; Fbx18; FLJ14590; hFBH1; MGC131916; MGC141935; MGC141937
Gene Aliases: FBH1; FBX18; FBXO18; hFBH1
UniProt ID: (Human) Q8NFZ0
Entrez Gene ID: (Human) 84893
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.