Proteintech
Manufacturer Code:142751AP150UL
Catalog # 50-172-9336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FBXO18 Fusion Protein Ag5622 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F box DNA helicase 1 F box only protein 18 F box protein helicase 18 FBH1 Fbx18 FBXO18; F-box DNA helicase 1; F-box only protein 18; F-box only protein, helicase, 18; hFBH1
Gene Aliases: AU015756; FBH1; FBX18; FBXO18; hFBH1
UniProt ID: (Human) Q8NFZ0, (Mouse) Q8K2I9
Entrez Gene ID: (Human) 84893, (Mouse) 50755, (Rat) 291293
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.