Invitrogen
Manufacturer Code:PA569228
Catalog # PIPA569228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human FBXO11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F-box only protein 11; F-box protein 11 FBX11 FLJ12673 MGC44383 PRMT9 UG063H01 VIT1 UBR6; GENA 104; jeff; Protein arginine N-methyltransferase 9; ubiquitin protein ligase E3 component n-recognin 6; VIT-1; Vitiligo-associated protein 1; vitiligo-associated protein VIT-1
Gene Aliases: C80048; FBX11; FBXO11; Jf; PRMT9; UBR6; UG063H01; VIT1
UniProt ID: (Human) Q86XK2, (Rat) Q7TSL3, (Mouse) Q7TPD1
Entrez Gene ID: (Horse) 100053097, (Dog) 481363, (Human) 80204, (Rabbit) 100351392, (Rat) 301674, (Mouse) 225055, (Guinea pig) 100729179
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.