Abnova
Manufacturer Code:H00080204A01
Catalog # 89-107-366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 744 a.a. to 843 a.a) partial recombinant protein with GST tag.,FBXO11 (NP_079409 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant FBXO11.
For Research Use Only
Protein Aliases: F-box only protein 11; FBX11; FLJ12673; MGC44383; PRMT9; Protein arginine N-methyltransferase 9; ubiquitin protein ligase E3 component n-recognin 6; UBR6; UG063H01; VIT-1; VIT1; Vitiligo-associated protein 1; vitiligo-associated protein VIT-1
Gene Aliases: FBX11; FBXO11; PRMT9; UBR6; UG063H01; VIT1
UniProt ID: (Human) Q86XK2
Entrez Gene ID: (Human) 80204
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.